Anti-IgG Donkey Polyclonal Antibody (Gold, 6nm)
Catalog # USBII1903-80B
Supplier: US Biological
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Gold, 6nm
- ELISA:Yes
- Host:Donkey
- Isotype:IgG
- Reactivity:Chicken
- Western blot:Yes
- Form:liquid
- Storage buffer:0,01M sodium borate-sodium phosphate, 0,15Msodium chloride, pH 8.5, 15mg/ml BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:300 µl
- Pk:300 µl
Specifications
About this item
Recommended dilution: ELISA: 1:20-1:40
Do not freeze!
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: 6 nm Gold
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Chicken