Anti-HTLV-1 gag p19 Mouse Monoclonal Antibody [clone: 8.A.024]
USBIH7950-32-50
- Antibody type:Primary
- Antigen symbol:HTLV-1 gag p19
- Clonality:Monoclonal
- Clone:8.A.024
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: HTLV-1 gag p19
Clonality: monoclonal
Clone: 8.A.024
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human