Anti-HLA Class 1 Antigen G Mouse Monoclonal Antibody [clone: MEM-G/9]
USBIH6098-72R
- Antibody type:Primary
- Antigen name:HLA Class 1 Antigen G
- Clonality:Monoclonal
- Clone:MEM-G/9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HLA Class 1 Antigen G
Clonality: monoclonal
Clone: MEM-G/9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human