Anti-HIV-1 gp41 Rabbit polyclonal antibody
Catalog # USBIH6004-12
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:HIV-1 gp41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: ELISA: 1:1000
Type: Primary
Antigen: HIV-1 gp41
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: