Order Entry
Norway
ContactUsLinkComponent
 
Anti-Histone H3, dimethyl K9 Mouse monoclonal antibody [clone: 9F619]
  USBIH5110-23N
Anti-Histone H3, dimethyl K9 Mouse monoclonal antibody [clone: 9F619]
  USBIH5110-23N

 

  • Antistofftype:
    Primary
  • Antigen-navn:
    Histone H3
  • Antigensymbol:
    H3
  • Klonalitet:
    Monoclonal
  • Klon:
    9F619
  • Konjugering:
    Unconjugated
  • ELISA:
    Yes
  • Vert:
    Mouse
  • Isotype:
    IgG1
  • Reaktivitet:
    Human
  • Skjema:
    liquid
  • Aminosyretall:
    4 to 14
  • Modifikasjon:
    dimethyl-Lys9
  • Lagringsbuffer:
    PBS, pH 7,2.
  • Lagringstemp:
    Cool
  • Frakttemp.:
    Ice
  • Rensing:
    purified by Protein G chromatography
  • Str:
    150 µg
  • Fp:
    150 µG

 

 

Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: Histone H3, dimethyl K9
Clonality: monoclonal
Clone: 9F619
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human