Anti-FOLR3 Mouse monoclonal antibody [clone: 9j7]
Catalog # USBIF5750-18
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Folate receptor 3
- Antigen symbol:FOLR3
- Clonality:Monoclonal
- Clone:9j7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:22 to 245
- Storage buffer:PBS, pH 7,4, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 2?g/ml
Type: Primary
Antigen: FOLR3 (folate receptor 3 (gamma))
Clonality: monoclonal
Clone: 9j7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human