Anti-FLT3LG Rabbit Polyclonal Antibody (Biotin)
USBIF4600-10G
- Antibody type:Primary
- Antigen name:Fms-related tyrosine kinase 3 ligand
- Antigen symbol:FLT3LG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: ELISA: 0.25-1 ?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: FLT3LG (fms-related tyrosine kinase 3 ligand)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human