Anti-EPHB2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K86]
Catalog # USBIE3371-04E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:EPH receptor B2
- Antigen symbol:EPHB2
- Clonality:Monoclonal
- Clone:12K86
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Form:liquid
- Amino acid number:27 to 548
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: EPHB2 (EPH receptor B2)
Clonality: monoclonal
Clone: 12K86
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse