Order Entry
Norway
ContactUsLinkComponent
 
Anti-Cytokeratin 5,8 Mouse Monoclonal Antibody [clone: 5F295]
  USBIC9097-37H1
Anti-Cytokeratin 5,8 Mouse Monoclonal Antibody [clone: 5F295]
  USBIC9097-37H1

 

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 5,8
  • Clonality:
    Monoclonal
  • Clone:
    5F295
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Hamster,
    Pig,
    Rabbit,
    Mammals,
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    Cell Culture supernatant
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Recommended dilution: Immunochemistry: 1:5-1:10

Type: Primary
Antigen: Cytokeratin 5,8
Clonality: Monoclonal
Clone: 5F295
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Hamster, Human, Mammalian, Mouse, Pig, Rabbit, Rat