Anti-CAP-G2 Rabbit Polyclonal Antibody
Catalog # USBIC1075-98F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CAP-G2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1100 to 1143
- Storage buffer:Tris saline, 0,1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:2000-1:10000; Immunoprecipitation: 1-4?g/mg
Type: Primary
Antigen: CAP-G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human