Anti-CNR2 Rabbit Polyclonal Antibody
USBIC1075-75N
- Antibody type:Primary
- Antigen name:cannabinoid receptor 2 (macrophage)
- Antigen symbol:CNR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 33
- Storage buffer:PBS, 1mg/ml BSA, 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Flow Cytometry: 1:50; Immunochemistry: 1:100;; Western Blot: 1:250
Type: Primary
Antigen: CNR2 (cannabinoid receptor 2 (macrophage))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat