Anti-BrdU Sheep polyclonal antibody unconjugated
Supplier: US Biological
Certificates
About this item
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG2b
Reactivity:
Specifications
- Catalog No:
- USBIB2850-01G-250
- USBIB2850-01G-50
- Antigen symbol:
- BrdU
- BrdU
- Antigen name:
- Bromodeoxyuridine
- Bromodeoxyuridine
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Sheep
- Sheep
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice