Anti-ATP5B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
USBIA3999-67-HRP
- Antibody type:Primary
- Antigen name:ATP Synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
- Antigen symbol:ATP5B
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:142 to 171
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Flow Cytometry: 1:10-1:50; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: ATP5B (ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human