Anti-ROR alpha Rabbit Polyclonal Antibody
USBI138300
- Antibody type:Primary
- Antigen symbol:ROR alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1. 25 ?g/ml; Immunochemistry: 4-8?g/ml
Type: Primary
Antigen: ROR alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat, Zebrafish