Order Entry
Norway
ContactUsLinkComponent
Anti-Matrix Metalloproteinase 3 Rabbit Polyclonal Antibody
Anti-Matrix Metalloproteinase 3 Rabbit Polyclonal Antibody
Catalog # USBI136925
Supplier:  US Biological
Anti-Matrix Metalloproteinase 3 Rabbit Polyclonal Antibody
Catalog # USBI136925
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 3
  • Antigen symbol:
    MMP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Matrix Metalloproteinase 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat