Order Entry
Norway
ContactUsLinkComponent
Anti-Dengue Virus, NS1 Mouse Monoclonal Antibody [clone: 13F45]
Anti-Dengue Virus, NS1 Mouse Monoclonal Antibody [clone: 13F45]
Catalog # USBI136587
Supplier:  US Biological
Anti-Dengue Virus, NS1 Mouse Monoclonal Antibody [clone: 13F45]
Catalog # USBI136587
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dengue Virus, NS1
  • Clonality:
    Monoclonal
  • Clone:
    13F45
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    0,1MPBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Dengue Virus, NS1
Clonality: monoclonal
Clone: 13F45
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse