Anti-SMAD7 Mouse Monoclonal Antibody [clone: 4E1]
Catalog # USBI133546
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:SMAD family member 7
- Antigen symbol:SMAD7
- Clonality:Monoclonal
- Clone:4E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:160 to 260
- Storage buffer:PBS, pH 7,2.
- Sequence:CKVFRWPDLRHSSEVKRLCCCESYGKINPELVCCNPHHLSRLCELESPPPPYSRYPMDFLKPTADCPDAVPSSAETGGTNYLAPGGLSDSQLLLEPGDRSH
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SMAD7 (SMAD family member 7)
Clonality: monoclonal
Clone: 40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human