Anti-ROR alpha Mouse Monoclonal Antibody [clone: 4E3]
Catalog # USBI132727
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:ROR alpha
- Clonality:Monoclonal
- Clone:4E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Amino acid number:424 to 524
- Storage buffer:PBS, pH 7,2.
- Sequence:SAFVLMSADRSWLQEKVKIEKLQQKIQLALQHVLQKNHREDGILTKLICKVSTLRALCGRHTEKLMAFKAIYPDIVRLHFPPLYKELFTSEFEPAMQIDG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ROR alpha
Clonality: monoclonal
Clone: 4000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human