Anti-POMT2 Mouse Monoclonal Antibody [clone: 1D9]
USBI131609
- Antibody type:Primary
- Antigen name:protein-O-mannosyltransferase 2
- Antigen symbol:POMT2
- Clonality:Monoclonal
- Clone:1D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:483 to 581
- Storage buffer:PBS, pH 7,2.
- Sequence:CVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFS*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: POMT2 (protein-O-mannosyltransferase 2)
Clonality: monoclonal
Clone: 1D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human