Anti-GPRC5D Mouse Monoclonal Antibody [clone: 6D9]
Catalog # USBI127506
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor, Class C, Group 5, Member D
- Antigen symbol:GPRC5D
- Clonality:Monoclonal
- Clone:6D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:261 to 346
- Storage buffer:PBS, pH 7,2.
- Sequence:ELCILYRSCRQECPLQGNACPVTAYQHSFQVENQELSRARDSDGAEEDVALTSYGTPIQPQTVDPTQECFIPQAKLSPQQDAGGV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GPRC5D (G protein-coupled receptor, class C, group 5, member D)
Clonality: monoclonal
Clone: 6D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human