Anti-GLYAT Mouse Monoclonal Antibody [clone: 1A10]
Catalog # USBI127375
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glycine-N-acyltransferase
- Antigen symbol:GLYAT
- Clonality:Monoclonal
- Clone:1A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:64 to 163
- Storage buffer:PBS, pH 7,2.
- Sequence:DMTDDLDHYTNTYQIYSKDPQNCQEFLGSPELINWKQHLQIQSSQPSLNEAIQNLAAIKSFKVKQTQRILYMAAETAKELTPFLLKSKILSPSGGKPKAI
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: GLYAT (glycine-N-acyltransferase)
Clonality: monoclonal
Clone: 1A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human