Anti-FAIM3 Mouse Monoclonal Antibody [clone: 1E4]
USBI126581
- Antibody type:Primary
- Antigen name:Fas apoptotic inhibitory molecule 3
- Antigen symbol:FAIM3
- Clonality:Monoclonal
- Clone:1E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:SEYEPSWEEQPMPETPKWFHLPYLFQMPAYASSSKFVTRVTTPAQRGKVPPVHHSSPTTQITHRPRVSRASSVAGDKPRTFLPSTTASKISALEGLLKPQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: FAIM3 (Fas apoptotic inhibitory molecule 3)
Clonality: monoclonal
Clone: 10000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human