Anti-ATG3 Mouse Monoclonal Antibody [clone: 1F7]
USBI123678
- Antibody type:Primary
- Antigen name:autophagy related 3
- Antigen symbol:ATG3
- Clonality:Monoclonal
- Clone:1F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 101
- Storage buffer:PBS, pH 7,2.
- Sequence:MQNVINTVKGKALEVAEYLTPVLKESKFKETGVITPEEFVAAGDHLVHHCPTWQWATGEELKVKAYLPTGKQFLVTKNVPCYKRCKQMEYSDELEAIIEE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: ATG3 (autophagy related 3)
Clonality: monoclonal
Clone: 1F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human