Anti-YME1L1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI043961-AP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:YME1-like 1 ATPase
- Antigen symbol:YME1L1
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Hamster,Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:140 to 170
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: YME1L1 (YME1-like 1 ATPase)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human