Anti-SLC12A7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
USBI041794-HRP
- Antibody type:Primary
- Antigen name:solute carrier family 12 (potassium/chloride transporter), member 7
- Antigen symbol:Solute carrier family 12, member 7
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:96 to 125
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: SLC12A7 (solute carrier family 12 (potassium/chloride transporter), member 7)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human