Anti-NAA60 Rabbit Polyclonal Antibody (Biotin)
USBI038807-BIOTIN
- Antibody type:Primary
- Antigen name:N(alpha)-acetyltransferase 60, NatF catalytic subunit
- Antigen symbol:NAA60
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:132 to 160
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: NAA60 (N(alpha)-acetyltransferase 60, NatF catalytic subunit)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse