Order Entry
Norway
ContactUsLinkComponent
Anti-LYSMD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-LYSMD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI038035-HRP
Supplier:  US Biological
undefined
Anti-LYSMD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI038035-HRP
Supplier:  US Biological

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    LysM, putative peptidoglycan-binding, domain containing 1
  • Antigensymbol:
    LYSMD1
  • Klonalitet:
    Polyclonal
  • Konjugering:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Vert:
    Rabbit
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Skjema:
    liquid
  • Aminosyretall:
    205 to 235
  • Lagringsbuffer:
    PBS, pH 7,2
  • Lagringstemp:
    Cool
  • Frakttemp.:
    Ice
  • Rensing:
    purified by Protein A chromatography
  • Str:
    200 µl
  • Fp:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: LYSMD1 (LysM, putative peptidoglycan-binding, domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse