Anti-COPE Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI034115-BIOTIN
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:coatomer Protein complex, subunit epsilon
- Antigen symbol:COPE
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:280 to 308
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50
Type: Primary
Antigen: COPE (coatomer protein complex, subunit epsilon)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human