Order Entry
Norway
ContactUsLinkComponent
 
Anti-CALCR Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI033146-APC
Anti-CALCR Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI033146-APC

 

  • Antistofftype:
    Primary
  • Antigen-navn:
    calcitonin receptor
  • Antigensymbol:
    CALCR
  • Klonalitet:
    Polyclonal
  • Konjugering:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flowcytometri:
    Yes
  • Vert:
    Rabbit
  • ImmunoFluorescens:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Skjema:
    liquid
  • Lagringsbuffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Lagringstemp:
    Cool
  • Frakttemp.:
    Ice
  • Rensing:
    purified by Protein A chromatography
  • Str:
    200 µl
  • Fp:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CALCR (calcitonin receptor)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human