Anti-ATP5C1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI032280-PE
- Antibody type:Primary
- Antigen name:ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1
- Antigen symbol:ATP5C1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:46 to 76
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: ATP5C1 (ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human