Anti-APOBEC3D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI031982-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3D
- Antigen symbol:APOBEC3D
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: APOBEC3D (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human