Anti-Lysosome-associated Membrane Protein 1 Mouse Monoclonal Antibody [clone: 13B830]
USBI031384
- Antibody type:Primary
- Antigen name:Lysosome Associated Membrane Protein 3
- Antigen symbol:LAMP3
- Clonality:Monoclonal
- Clone:13B830
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 10mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Immunofluorescence: 1:2000; Western Blot: 1:10000
Type: Primary
Antigen: Lysosome-associated Membrane Protein 1
Clonality: Monoclonal
Clone: 13B830
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Pig, Rat