Anti-MLST8 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 211CT7.1.1]
USBI030985-HRP
- Antibody type:Primary
- Antigen name:MTOR associated Protein, LST8 homolog (S. cerevisiae)
- Antigen symbol:MLST8
- Clonality:Monoclonal
- Clone:211CT7.1.1
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1560
Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: Monoclonal
Clone: 211CT7.1.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse