Order Entry
Norway
ContactUsLinkComponent
 
Anti-5-Methylcytosine Mouse Monoclonal Antibody [clone: 13A149]
  USBI029406
Anti-5-Methylcytosine Mouse Monoclonal Antibody [clone: 13A149]
  USBI029406

 

  • Antibody type:
    Primary
  • Antigen name:
    5-Methylcytosine
  • Clonality:
    Monoclonal
  • Clone:
    13A149
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 lambda
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Dot Blot: 1 ?g/ml

Type: Primary
Antigen: 5-Methylcytosine
Clonality: Monoclonal
Clone: 13A149
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human