Anti-CD40L Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: MR1]
Catalog # ROCK200-B26-N91
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:CD40 ligand
- Antigen symbol:CD40LG
- Clonality:Monoclonal
- Clone:MR1
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Armenian hamster
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:21947
- Antigen synonyms:TRAP|IMD3|gp39|CD154|CD40-L|Cd40l|Ly-62|IGM|T-BAM|Ly62|Tnfsf5|HIGM1
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,2 mg/ml Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD154 Antibody (Monoclonal) was produced by repeated immunizations with Activated mouse Th1 clone D1.6.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-CD154 is useful for immunochemistry and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: MR1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse