Order Entry
Norway
ContactUsLinkComponent
Anti-CD69 Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: H1.2F3]
Anti-CD69 Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: H1.2F3]
  ROCK200-B08-N88
undefined
Anti-CD69 Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: H1.2F3]
  ROCK200-B08-N88

 

  • Antibody type:
    Primary
  • Antigen name:
    CD69 antigen
  • Antigen symbol:
    CD69
  • Clonality:
    Monoclonal
  • Clone:
    H1.2F3
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Armenian hamster
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    12515
  • Antigen synonyms:
    5830438K24Rik|AIM|VEA|AI452015
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    0,2 mg/ml Sufficient to run approximately 100 tests
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-CD69 Antibody (Monoclonal) was produced by repeated immunizations with Mouse dendritic epidermal T cell line Y245.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    200 µg
  • Pk:
    200 µG

 

 

Anti-CD69 is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse