Order Entry
Norway
ContactUsLinkComponent
Anti-CD19 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 6D5]
Anti-CD19 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 6D5]
Catalog # ROCK200-502-N81
undefined
Anti-CD19 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 6D5]
Catalog # ROCK200-502-N81

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD19 antigen
  • Antigen symbol:
    CD19
  • Clonality:
    Monoclonal
  • Clone:
    6D5
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    12478
  • Antigen synonyms:
    AW495831
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    Sufficient to run approximately 100 tests
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-CD19 Antibody (Monoclonal) was produced by repeated immunizations with Mouse CD19-expressing K562 human erythroleukemia cells.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Anti-CD19 is useful for Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse