Order Entry
Norway
ContactUsLinkComponent
 
Anti-KLRB1C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: PK136]
  ROCK200-326-N93
undefined
Anti-KLRB1C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: PK136]
  ROCK200-326-N93

 

  • Antibody type:
    Primary
  • Antigen name:
    killer cell lectin-like receptor subfamily B member 1C
  • Antigen symbol:
    KLRB1C
  • Clonality:
    Monoclonal
  • Clone:
    PK136
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    17059
  • Antigen synonyms:
    NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
  • Storage temperature:
    Cool
  • Concentration:
    0,2 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-NK1.1 Antibody (Monoclonal) was produced by repeated immunizations with NK1.1 antigen.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Anti-NK1.1 is useful for immunochemistry, Immunoprecipitation, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: KLRB1C
Clonality: Monoclonal
Clone: PK136
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse