Anti-CD86 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: IT2.2]
Catalog # ROCK200-308-N75
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:CD86 molecule
- Antigen symbol:CD86
- Clonality:Monoclonal
- Clone:IT2.2
- Conjugation:RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:942
- Antigen synonyms:B70|B7.2|CD28LG2|LAB72|B7-2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,2% BSA (w/v)
- Storage temperature:Cool
- Concentration:Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD86 Antibody (Monoclonal) was produced by repeated immunizations with CD86 antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Anti-CD86 is useful for Flow Cytometry, immunochemistry, and Western Blot. Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze! Light sensitive!
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: IT2.2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human