Anti-lambda light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # ROCK609-1311
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen name:lambda light chain
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Thimerosal, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-Human lambda chain was produced by repeated immunization with human lambda light chain in goat.
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Human lambda chain Antibody peroxidase conjugated has been assayed against 1.0 µg of Human IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human