Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK609-1206
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Human IgA alpha heavy chain
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Secondary Goat Anti-IgA (alpha chain) Reacts with Human
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Human