Order Entry
Norway
ContactUsLinkComponent
Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ROCK600-402-466
undefined
Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ROCK600-402-466

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Keyhole Limpet Hemocyanin
  • Antigen symbol:
    KLH
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Goat
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    KLH (Keyhole Limpet Hemocyanin
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: