Order Entry
Norway
ContactUsLinkComponent
 
Anti-ATG8 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-439
undefined
Anti-ATG8 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-439

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Autophagy related 8
  • Antigen symbol:
    ATG8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Yeast
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Gene ID:
    852200
  • Antigen synonyms:
    ATG8
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    5,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    This purified antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast APG8 Protein.
  • Purification:
    multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

This purified polyclonal antibody reacts with yeast APG8 by western blot and ELISA.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:4000 - 1:8000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: ATG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast