Anti-CSF2 Rabbit Monoclonal Antibody (Unconjugated) [clone: 7A11]
Catalog # ROCK109-401-314
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:colony stimulating factor 2 (granulocyte-macrophage)
- Antigen symbol:CSF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:1437
- Antigen synonyms:GMCSF
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with full length recombinant human GM-CSF.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
This antiserum against Human GM-CSF has been tested for use in neutralizations, ELISA and immunoblotting. Reactivity is also expected in neutralizations and radioimmunoassay.
Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 7A11
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human