Anti-TNFA Rabbit Polyclonal Antibody
Catalog # ROCK109-401-306
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor
- Antigen symbol:TNF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Primates,Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:7124
- Antigen synonyms:DIF|TNF-alpha|TNFSF2|TNFA
- Storage temperature:Cool
- Concentration:75 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:The whole rabbit serum was prepared by repeated immunizations with recombinant human TNFa produced in E.coli.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Primary Rabbit Anti-Human TNF alpha Reacts with Human
Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primate