Anti-ADH1 Rabbit Polyclonal Antibody
Katalog # ROCK100-4143
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Alcohol Dehydrogenase
- Antigen symbol:ADH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Yeast
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Gene ID:2538902
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with full length Alcohol Dehydrogenase isolated from yeast.
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Specifications
About this item
This antibody has been tested for use in ELISA and by western blot.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: ADH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Yeast