Order Entry
Norway
ContactUsLinkComponent
Anti-ANGPT1 Rabbit Polyclonal Antibody
Anti-ANGPT1 Rabbit Polyclonal Antibody
Certificates
undefined
Anti-ANGPT1 Rabbit Polyclonal Antibody

Specifications

  • Catalog No:
  • ROCK100-401-403
  • ROCK100-401-403S
  • Antigen symbol:
  • ANGPT1
  • ANGPT1
  • Antigen name:
  • angiopoietin 1
  • Angiopoietin 1
  • Antigen synonyms:
  • Ang1|Ang-1|1110046O21Rik
  • ANG1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 11600
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Epitope:
  • near the N-terminus
  • Immunogen:
  • This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide N-Q-R-R-N-P-E-N-G-G-R-R-Y-N-R-I-Q-H-G-Q corresponding to a region (aa 21-40) near the N-terminus of mouse angiopoietin 1 protein conjugated to KLH using maleimide. A r
  • Amino acid number:
  • 21 to 40
  • Sequence:
  • NQRRNPENGGRRYNRIQHGQ
  • Purification:
  • Antiopoietin 1 is directed against mouse angiopoietin1 and shows no reactivity with mouse angiopoietin2.  A 57 kDa band is detected when assayed by immunoblot.  This reagent cross-reacts with human angiopoietin 1.  Partially cross reactivity is noted with human angiopoietin 2.
  • Concentration:
  • 80 mg/ml by Refractometry
  • Cross adsorption:
  • none
  • Storage buffer:
  • 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications