Anti-PTGS2 Rabbit Polyclonal Antibody
Katalog # ROCK100-401-226
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Prostaglandin-endoperoxide Synthase 2
- Antigen symbol:PTGS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:5743
- Antigen synonyms:hCox-2|COX-2|PHS-2|GRIPGHS|COX2|PGG/HS|PGHS-2
- Storage buffer:0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:55 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with a fusion Protein corresponding to the carboxy-terminus of human Cox-2.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Cox-2 (Cyclooxygenase-2) is an inducible enzyme that is normally absent from cells, however, in response to growth factors, tumor promoters and some cytokines, it undergoes a rapid and transient expression.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:5000 - 1:10000;
Type: Primary
Antigen: PTGS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human