Order Entry
Norway
ContactUsLinkComponent
Anti-ORM1 Rabbit Polyclonal Antibody
Anti-ORM1 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-046
Anti-ORM1 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-046

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    orosomucoid 1
  • Antigensymbol:
    ORM1
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Unconjugated
  • ELISA:
    Yes
  • Vert:
    Rabbit
  • Immunutfelling:
    Yes
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Kyssabsorpsjon:
    none
  • Format:
    Antiserum
  • Skjema:
    Lyophilized
  • Gen-ID:
    5004
  • Antigen synonymer:
    AGP1|HEL-S-153w|ORM|AGP-A
  • Lagringsbuffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Lagringstemp:
    Cool
  • Konsentrasjon:
    90 mg/ml by Refractometry
  • Frakttemp.:
    Ice
  • Immunogen:
    Alpha-1-Acid GlycoProtein (Human Plasma)
  • Rensing:
    delipidation and defibrination
  • Str:
    2 ml
  • Fp:
    2 mL

Specifications

About this item

This product has been assayed against 1.0 µg of Alpha-1-Acid GlycoProtein (Human Plasma) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human