Anti-ACHE Goat Polyclonal Antibody
ROCK100-101-232
- Antibody type:Primary
- Antigen name:Acetylcholinesterase
- Antigen symbol:ACHE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Reactivity:Bovine
- Western blot:Yes
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Gene ID:540446
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Acetyl Cholinesterase (Bovine Erythrocytes)
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
This product has been assayed against 1.0 µg of Acetyl Cholinesterase (Bovine Erythrocytes) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine